Learn More
Methyl Propionate, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 149052500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl propionate | |
554-12-1 | |
-88°C | |
0.9150g/mL | |
-2°C | |
99% min. (GC) | |
C4H8O2 | |
C2H5CO2CH3 | |
250 mL | |
0.915 | |
methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural | |
RJUFJBKOKNCXHH-UHFFFAOYSA-N | |
methyl propanoate | |
11124 | |
99+% |
99+% | |
1mg KOH/g max. | |
Colorless | |
78°C to 79°C | |
Authentic | |
Glass bottle | |
1.377 | |
MFCD00009306 | |
02, 239 | |
15, 6185 | |
Solubility in water: 5g/L water (20°C). Other solubilities: miscible with alcohol and ether,soluble in most common organic solvents | |
CCC(=O)OC | |
88.11 | |
88.11 | |
Liquid |
Chemical Identifiers
554-12-1 | |
88.11 | |
RJUFJBKOKNCXHH-UHFFFAOYSA-N | |
11124 | |
CCC(=O)OC |
C4H8O2 | |
MFCD00009306 | |
methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural | |
methyl propanoate |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Highly flammable liquid and vapor.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-060-4
RTECSNumber : UF5970000
TSCA : TSCA
RUO – Research Use Only