missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl Propargyl Ether 98.0+%, TCI America™
Supplier: TCI America M086025ML
Specifications
Methyl Propargyl Ether | |
Yellow | |
C4H6O | |
25 mL | |
Methyl 2-Propynyl Ether | |
COCC#C | |
70.09 | |
70.09 | |
Liquid |
627-41-8 | |
62°C | |
MFCD00008578 | |
3271 | |
YACFFSVYSPMSGS-UHFFFAOYSA-N | |
3-methoxyprop-1-yne | |
69393 | |
≥98.0% (GC) |
Chemical Identifiers
627-41-8 | |
70.09 | |
YACFFSVYSPMSGS-UHFFFAOYSA-N | |
69393 | |
COCC#C |
C4H6O | |
MFCD00008578 | |
Methyl 2-Propynyl Ether | |
3-methoxyprop-1-yne |
Safety and Handling
TSCA : No