Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl piperidine-4-carboxylate, 98%

Chemical Identifiers

Chemical Name or Material Methyl piperidine-4-carboxylate
CAS 2971-79-1
Formula Weight 143.18
MDL Number MFCD00190578
Beilstein 224128
Synonym Piperidine-4-carboxylic acid methyl ester; Methyl isonipecotate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6175414

 
alfa aesar™
H6175414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6175422

 
alfa aesar™
H6175422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 143.18
Boiling Point 85° to 90°C
Chemical Name or Material Methyl piperidine-4-carboxylate
Refractive Index 1.4650
Physical Form Liquid
Assay 98%
MDL Number MFCD00190578
CAS 2971-79-1
Beilstein 224128
Synonym Piperidine-4-carboxylic acid methyl ester; Methyl isonipecotate
Color Colorless to pale yellow
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.06g/mL
Quantity 25g

Applications
Methyl piperidine-4-carboxylate is a reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activation. It is used as a reactant for synthesis of antitubercular agents, aminopyrazine inhibitors, orally available naphthyridine protein kinase D inhibitors and a scalable catalyst from asymmetric hydrogenation of sterically demanding aryl enamide.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.