Learn More
Methyl picolinate, 99%, Thermo Scientific Chemicals
$211.44 - $570.99
Chemical Identifiers
CAS | 2459-07-6 |
---|---|
Molecular Formula | C7H7NO2 |
Molecular Weight (g/mol) | 137.14 |
MDL Number | MFCD00038038 |
InChI Key | NMMIHXMBOZYNET-UHFFFAOYSA-N |
Synonym | methyl picolinate, 2-picolinic acid methyl ester, 2-pyridinecarboxylic acid, methyl ester, methyl 2-pyridinecarboxylate, picolinic acid, methyl ester, 2-carbomethoxypyridine, pyridine-2-carboxylic acid methyl ester, 2-pyridinecarboxylic acid methyl ester, picolinic acid methyl ester, methylpicolinate |
PubChem CID | 75569 |
IUPAC Name | methyl pyridine-2-carboxylate |
SMILES | COC(=O)C1=CC=CC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5453014
|
Thermo Scientific Chemicals
H5453014 |
25 g |
Each for $211.44
|
|
|||||
AAH5453022
|
Thermo Scientific Chemicals
H5453022 |
100 g |
Each for $570.99
|
|
|||||
Description
Methyl picolinate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl picolinate is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
2459-07-6 | |
137.14 | |
NMMIHXMBOZYNET-UHFFFAOYSA-N | |
75569 | |
COC(=O)C1=CC=CC=N1 |
C7H7NO2 | |
MFCD00038038 | |
methyl picolinate, 2-picolinic acid methyl ester, 2-pyridinecarboxylic acid, methyl ester, methyl 2-pyridinecarboxylate, picolinic acid, methyl ester, 2-carbomethoxypyridine, pyridine-2-carboxylic acid methyl ester, 2-pyridinecarboxylic acid methyl ester, picolinic acid methyl ester, methylpicolinate | |
methyl pyridine-2-carboxylate |
Specifications
2459-07-6 | |
1.137 | |
103°C (217°F) | |
1.521 | |
25 g | |
Slightly soluble in water. | |
COC(=O)C1=CC=CC=N1 | |
137.14 | |
137.14 | |
Methyl picolinate |
19°C | |
95°C (1 mmHg) | |
C7H7NO2 | |
MFCD00038038 | |
methyl picolinate, 2-picolinic acid methyl ester, 2-pyridinecarboxylic acid, methyl ester, methyl 2-pyridinecarboxylate, picolinic acid, methyl ester, 2-carbomethoxypyridine, pyridine-2-carboxylic acid methyl ester, 2-pyridinecarboxylic acid methyl ester, picolinic acid methyl ester, methylpicolinate | |
NMMIHXMBOZYNET-UHFFFAOYSA-N | |
methyl pyridine-2-carboxylate | |
75569 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 219-545-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only