Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl (phenylthio)acetate, 99%

$44.00 - $137.12

Chemical Identifiers

Chemical Name or Material Methyl (phenylthio)acetate
CAS 17277-58-6
Molecular Formula C6H5SCH2CO2CH3
Formula Weight 182.24
MDL Number MFCD00039805
Beilstein 2047554
Synonym Methyl (phenylmercapto)acetate; (Phenylthio)acetic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1264906

  SDS
alfa aesar™
L1264906
5g Each for $44.00

AAL1264914

  SDS
alfa aesar™
L1264914
25g Each for $137.12
Description & Specifications

Specifications

Formula Weight 182.24
Chemical Name or Material Methyl (phenylthio)acetate
Assay 99%
Molecular Formula C6H5SCH2CO2CH3
MDL Number MFCD00039805
CAS 17277-58-6
Synonym Methyl (phenylmercapto)acetate; (Phenylthio)acetic acid methyl ester
Health Hazard 1 Warning
Boiling Point 104° to 105°C (4mmHg)
Flash Point >110°C (230°F)
Odor Stench
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2047554
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.