missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl (phenylthio)acetate, 99%

Manufacturer:  Alfa Aesar™ L1264914

 View more versions of this product

Catalog No. AAL1264914



Specifications

Specifications

Methyl (phenylthio)acetate
Stench
25g
C9H10O2S
2047554
MUNSXQQODXYRKI-UHFFFAOYSA-N
methyl 2-phenylsulfanylacetate
123375
99%
>110°C (230°F)
104°C to 105°C (4mmHg)
17277-58-6
MFCD00039805
methyl phenylthio acetate, methyl phenylthio acetate, methyl 2-phenylthio acetate, methyl phenylsulfanyl acetate, phenylthio acetic acid methyl ester, methyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, methyl ester, methyl 2-phenylthioacetate, methyl phenylsulfanyl acetate, pubchem10851
COC(=O)CSC1=CC=CC=C1
182.237
182.24

Chemical Identifiers

17277-58-6
182.237
MUNSXQQODXYRKI-UHFFFAOYSA-N
123375
COC(=O)CSC1=CC=CC=C1
C9H10O2S
MFCD00039805
methyl phenylthio acetate, methyl phenylthio acetate, methyl 2-phenylthio acetate, methyl phenylsulfanyl acetate, phenylthio acetic acid methyl ester, methyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, methyl ester, methyl 2-phenylthioacetate, methyl phenylsulfanyl acetate, pubchem10851
methyl 2-phenylsulfanylacetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents