Learn More
Methyl phenylsulfonylacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0374814
Description
Methyl phenylsulfonylacetate has been used in a key step during the synthesis of (-)-hybridalactone, marine eicosanoid isolated from the red alga Laurencia hybrida, in dehydrative alkylation of alcohols under modified Mitsunobu conditions followed by desulfonylation using magnesium, in three step synthesis of 2-(hydroxymethyl)indene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl phenylsulfonylacetate has been used in a key step during the synthesis of (-)-hybridalactone, marine eicosanoid isolated from the red alga Laurencia hybrida, in dehydrative alkylation of alcohols under modified Mitsunobu conditions followed by desulfonylation using magnesium, in three step synthesis of 2-(hydroxymethyl)indene.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Methyl phenylsulfonylacetate | |
∼30°C | |
165°C (0.05 mmHg) | |
C9H10O4S | |
MFCD00007555 | |
2051463 | |
Insoluble in water. | |
COC(=O)CS(=O)(=O)C1=CC=CC=C1 | |
214.235 | |
214.24 |
34097-60-4 | |
1.24 | |
>110°C (230°F) | |
1.535 | |
25 g | |
methyl phenylsulfonylacetate, methyl 2-phenylsulfonyl acetate, methyl phenylsulfonyl acetate, methylphenylsulfonylacetate, benzenesulfonyl-acetic acid methyl ester, acmc-1ad0h, methyl benzenesulfonylacetate, methyl2-phenylsulfonyl acetate, methyl-phenylsulfonyl acetate | |
NLEAIFBNKPYTGN-UHFFFAOYSA-N | |
methyl 2-(benzenesulfonyl)acetate | |
555619 | |
98% |
Chemical Identifiers
34097-60-4 | |
214.235 | |
NLEAIFBNKPYTGN-UHFFFAOYSA-N | |
methyl 2-(benzenesulfonyl)acetate |
C9H10O4S | |
MFCD00007555 | |
555619 | |
COC(=O)CS(=O)(=O)C1=CC=CC=C1 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only