missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Phenylpropiolate 98.0+%, TCI America™
$164.03 - $568.54
Chemical Identifiers
| CAS | 4891-38-7 |
|---|---|
| Molecular Formula | C10H8O2 |
| Molecular Weight (g/mol) | 160.172 |
| MDL Number | MFCD00041685 |
| InChI Key | JFGWPXKGINUNDH-UHFFFAOYSA-N |
| Synonym | methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate |
| PubChem CID | 138378 |
| IUPAC Name | methyl 3-phenylprop-2-ynoate |
| SMILES | COC(=O)C#CC1=CC=CC=C1 |
Chemical Identifiers
| 4891-38-7 | |
| 160.172 | |
| JFGWPXKGINUNDH-UHFFFAOYSA-N | |
| 138378 | |
| COC(=O)C#CC1=CC=CC=C1 |
| C10H8O2 | |
| MFCD00041685 | |
| methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate | |
| methyl 3-phenylprop-2-ynoate |
Specifications
| 4891-38-7 | |
| 103°C | |
| MFCD00041685 | |
| methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate | |
| COC(=O)C#CC1=CC=CC=C1 | |
| 160.172 | |
| 160.17 | |
| Crystalline Lumps |
| White-Yellow | |
| C10H8O2 | |
| 5 g | |
| JFGWPXKGINUNDH-UHFFFAOYSA-N | |
| methyl 3-phenylprop-2-ynoate | |
| 138378 | |
| ≥98.0% (GC) | |
| Methyl Phenylpropiolate |
Safety and Handling
TSCA : No