Please login to your online account to display your discounted pricing

Methyl phenylpropiolate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  334590250

 View more versions of this product

Catalog No. AC334590250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid After Melting
Formula Weight 160.17
Quantity 25g
Color Clear colorless to pale yellow
Chemical Name or Material Methyl phenylpropiolate
Refractive Index 1.5620 to 1.5640 (20°C, 589nm)
MDL Number MFCD00041685
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10H8O2
CAS 4891-38-7

Specifications

Chemical Name or Material Methyl phenylpropiolate
Name Note 98%
CAS 4891-38-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H8O2
Formula Weight 160.17
MDL Number MFCD00041685