Please login to your online account to display your discounted pricing

Methyl phenylpropiolate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  334590050

 View more versions of this product

Catalog No. AC334590050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 160.17
Physical Form Liquid After Melting
Quantity 5g
Color Clear colorless to pale yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5620 to 1.5640 (20°C, 589nm)
CAS 4891-38-7
MDL Number MFCD00041685
Name Note 98%
Molecular Formula C10H8O2
Assay Percent Range 98%
Chemical Name or Material Methyl phenylpropiolate

Specifications

Chemical Name or Material Methyl phenylpropiolate
Name Note 98%
CAS 4891-38-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H8O2
Formula Weight 160.17
MDL Number MFCD00041685