missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl phenylpropiolate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2458722
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl phenylpropiolate | |
1.083 | |
>110°C (230°F) | |
C6H5C≡CCO2CH3 | |
MFCD00041685 | |
472813 | |
JFGWPXKGINUNDH-UHFFFAOYSA-N | |
methyl 3-phenylprop-2-ynoate | |
138378 | |
97% |
4891-38-7 | |
109°C to 110°C (2 mmHg) | |
C10H8O2 | |
1.562 | |
100 g | |
methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate | |
COC(=O)C#CC1=CC=CC=C1 | |
160.172 | |
160.17 |
Chemical Identifiers
4891-38-7 | |
160.172 | |
JFGWPXKGINUNDH-UHFFFAOYSA-N | |
138378 | |
COC(=O)C#CC1=CC=CC=C1 |
C10H8O2 | |
MFCD00041685 | |
methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate | |
methyl 3-phenylprop-2-ynoate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only