missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl phenyl sulfoxide, 98+%

Manufacturer:  Alfa Aesar™ A1500922

 View more versions of this product

Catalog No. AAA1500922



Specifications

Specifications

Methyl phenyl sulfoxide
93°C to 94°C (0.3mmHg)
100g
C7H8OS
1904976
JXTGICXCHWMCPM-UHFFFAOYSA-N
methylsulfinylbenzene
14516
≥98%
86°C (186°F)
26°C to 32°C
1193-82-4
MFCD00002088
methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene, methylsulfinyl benzene
CS(=O)C1=CC=CC=C1
140.2
140.2

Chemical Identifiers

1193-82-4
140.2
JXTGICXCHWMCPM-UHFFFAOYSA-N
14516
CS(=O)C1=CC=CC=C1
C7H8OS
MFCD00002088
methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene, methylsulfinyl benzene
methylsulfinylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents