missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl phenyl sulfone, 98+%

Manufacturer:  Alfa Aesar™ A1172730

 View more versions of this product

Catalog No. AAA1172730



Description

Description

Applications
Methyl phenyl sulfone, a versatile building block for the construction of chemicals with diverse functionalities

Solubility
Soluble in methanol, ethanol, benzene, and chloroform. Partly miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Methyl phenyl sulfone
250g
C7H8O2S
1906914
JCDWETOKTFWTHA-UHFFFAOYSA-N
methylsulfonylbenzene
18369
≥98%
85°C to 88°C
3112-85-4
MFCD00014741
methyl phenyl sulfone, methylsulfonyl benzene, phenyl methyl sulfone, benzene, methylsulfonyl, sulfone, methyl phenyl, phenylsulfonyl methane, methylphenylsulfone, methylsulphonyl benzene, methanesulfonylbenzene, methyl phenyl sulphone
CS(=O)(=O)C1=CC=CC=C1
156.199
156.2

Chemical Identifiers

3112-85-4
156.199
JCDWETOKTFWTHA-UHFFFAOYSA-N
18369
CS(=O)(=O)C1=CC=CC=C1
C7H8O2S
MFCD00014741
methyl phenyl sulfone, methylsulfonyl benzene, phenyl methyl sulfone, benzene, methylsulfonyl, sulfone, methyl phenyl, phenylsulfonyl methane, methylphenylsulfone, methylsulphonyl benzene, methanesulfonylbenzene, methyl phenyl sulphone
methylsulfonylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : WR6800000

SDS Documents