missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl phenyl carbonate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2523206

 View more versions of this product

Catalog No. AAB2523206



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications

Specifications

Methyl phenyl carbonate
210°C to 213°C
1.521
5 g
XTBFPVLHGVYOQH-UHFFFAOYSA-N
methyl phenyl carbonate
139482
97%
13509-27-8
C8H8O3
MFCD04039782
carbonic acid, methyl phenyl ester, phenyl methyl carbonate, phenyl methoxyformate, acmc-20akf2, carbonic acid methyl phenyl
COC(=O)OC1=CC=CC=C1
152.15
152.15

Chemical Identifiers

13509-27-8
152.15
XTBFPVLHGVYOQH-UHFFFAOYSA-N
139482
COC(=O)OC1=CC=CC=C1
C8H8O3
MFCD04039782
carbonic acid, methyl phenyl ester, phenyl methyl carbonate, phenyl methoxyformate, acmc-20akf2, carbonic acid methyl phenyl
methyl phenyl carbonate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only