Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl perfluorotetradecanoate, 96%


Quantity 5g

Manufacturer: alfa aesar™  L1697506

 View more versions of this product

Catalog No. AAL1697506

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 728.15
Chemical Name or Material Methyl perfluorotetradecanoate
Assay 96%
Molecular Formula C15F27H3O2
Linear Formula CF3(CF2)12CO2CH3
MDL Number MFCD00236620
Synonym Perfluorodecanoic acid methyl ester
CAS 203302-99-2
Melting Point 95° to 98°C
Quantity 5g

Specifications

Chemical Name or Material Methyl perfluorotetradecanoate
CAS 203302-99-2
Molecular Formula C15F27H3O2
Linear Formula CF3(CF2)12CO2CH3
Formula Weight 728.15
MDL Number MFCD00236620
Synonym Perfluorodecanoic acid methyl ester