Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl perfluorotetradecanoate, 96%

$99.59 - $99.59

Chemical Identifiers

Chemical Name or Material Methyl perfluorotetradecanoate
CAS 203302-99-2
Molecular Formula C15F27H3O2
Linear Formula CF3(CF2)12CO2CH3
Formula Weight 728.15
MDL Number MFCD00236620
Synonym Perfluorodecanoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1697506

 
alfa aesar™
L1697506
5g Each for $99.59
Description & Specifications

Specifications

Formula Weight 728.15
Chemical Name or Material Methyl perfluorotetradecanoate
Assay 96%
Molecular Formula C15F27H3O2
Linear Formula CF3(CF2)12CO2CH3
MDL Number MFCD00236620
Synonym Perfluorodecanoic acid methyl ester
CAS 203302-99-2
Melting Point 95° to 98°C
Quantity 5g