Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl perfluorooctanoate, 97%

$37.81 - $37.81

Chemical Identifiers

Chemical Name or Material Methyl perfluorooctanoate
CAS 376-27-2
Molecular Formula CF3(CF2)6CO2CH3
Formula Weight 428.09
MDL Number MFCD00039244
Beilstein 1809817
Synonym Methyl pentadecafluorooctanotate; Perfluorooctanoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2385606

  SDS
alfa aesar™
B2385606
5g Each for $37.81
Description & Specifications

Specifications

Formula Weight 428.09
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 376-27-2
Chemical Name or Material Methyl perfluorooctanoate
Quantity 5g
Molecular Formula CF3(CF2)6CO2CH3
Beilstein 1809817
Flash Point 65°C (149°F)
TSCA TSCA
Boiling Point 158° to 159°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
MDL Number MFCD00039244
RTECS Number RH0783200
Assay 97%
Synonym Methyl pentadecafluorooctanotate; Perfluorooctanoic acid methyl ester
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.