Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl perfluorononanoate, 96%

$68.70 - $194.38

Chemical Identifiers

Chemical Name or Material Methyl perfluorononanoate
CAS 51502-45-5
Molecular Formula C10H3F17O2
Linear Formula CF3(CF2)7CO2CH3
Formula Weight 478.11
MDL Number MFCD00153213
Beilstein 2315727
Synonym Perfluorononanoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1681006

 
alfa aesar™
L1681006
5g Each for $68.70

AAL1681014

 
alfa aesar™
L1681014
25g Each for $194.38
Description & Specifications

Specifications

Formula Weight 478.11
MDL Number MFCD00153213
Chemical Name or Material Methyl perfluorononanoate
Assay 96%
Molecular Formula C10H3F17O2
Linear Formula CF3(CF2)7CO2CH3
CAS 51502-45-5
Beilstein 2315727
Synonym Perfluorononanoic acid methyl ester
Flash Point >110°C (230°F)
Boiling Point 63° to 65°C (10mmHg)
Quantity 5g