missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl perfluorododecanoate, 96%

$72.16 - $195.85

Chemical Identifiers

CAS 56554-52-0
Molecular Formula C13H3F23O2
Molecular Weight (g/mol) 628.128
MDL Number MFCD00236616
InChI Key ABNZIVVYHQYSLI-UHFFFAOYSA-N
Synonym methyl perfluorododecanoate, methyl tricosafluorododecanoate, tricosafluorododecanoic acid methyl ester, dodecanoic acid, tricosafluoro-, methyl ester, methyl tricosafluorolaurate, acmc-1b030, methyl perfluorododecanoate, perfluorododecanoic acid methyl ester, methyl tricosafluorolaurate; perfluorododecanoic acid methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid methyl ester
PubChem CID 547885
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoate
SMILES COC(=O)C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1680906 View Documents Alfa Aesar™
L1680906
5g Each for $72.16
 
AAL1680914 View Documents Alfa Aesar™
L1680914
25g Each for $195.85
 
Specifications

Chemical Identifiers

56554-52-0
628.128
ABNZIVVYHQYSLI-UHFFFAOYSA-N
547885
COC(=O)C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C13H3F23O2
MFCD00236616
methyl perfluorododecanoate, methyl tricosafluorododecanoate, tricosafluorododecanoic acid methyl ester, dodecanoic acid, tricosafluoro-, methyl ester, methyl tricosafluorolaurate, acmc-1b030, methyl perfluorododecanoate, perfluorododecanoic acid methyl ester, methyl tricosafluorolaurate; perfluorododecanoic acid methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid methyl ester
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoate

Specifications

54°C to 56°C
56554-52-0
CF3(CF2)10CO2CH3
methyl perfluorododecanoate, methyl tricosafluorododecanoate, tricosafluorododecanoic acid methyl ester, dodecanoic acid, tricosafluoro-, methyl ester, methyl tricosafluorolaurate, acmc-1b030, methyl perfluorododecanoate, perfluorododecanoic acid methyl ester, methyl tricosafluorolaurate; perfluorododecanoic acid methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid methyl ester
COC(=O)C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
628.128
628.13
Methyl perfluorododecanoate
5g
C13H3F23O2
MFCD00236616
ABNZIVVYHQYSLI-UHFFFAOYSA-N
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoate
547885
96%