missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl perfluoro-2,5-dimethyl-3,6-dioxanonanoate, 97%

$47.35 - $153.12

Chemical Identifiers

CAS 26131-32-8
Molecular Formula C10H3F17O4
Molecular Weight (g/mol) 510.103
MDL Number MFCD00054723
InChI Key UMADKHFVSXRTGY-UHFFFAOYSA-N
Synonym methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-perfluoropropoxy propoxy propanoate, perfluoro-2,5-dimethyl-3,6-dioxanonanoic acid methyl ester, methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propanoate, methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-heptafluoropropoxy propoxy propanoate, cf3cf2cf2ocf cf3 cf2ocf cf3 cooch3, methyl perfluoro-2,5-dimethyl-3,6-dioxa-nonanoate, methyl 2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoate, 2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoic acid methyl ester, 2-2-heptafluoropropoxy hexafluoropropoxy-2,3,3,3-tetrafluoropropionic acid methyl ester, methyl2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoate mixtureofisomers
PubChem CID 141268
IUPAC Name methyl 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoate
SMILES COC(=O)C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1658603 View Documents Alfa Aesar™
L1658603
1g Each for $47.35
 
AAL1658606 View Documents Alfa Aesar™
L1658606
5g Each for $153.12
 
Specifications

Chemical Identifiers

26131-32-8
510.103
UMADKHFVSXRTGY-UHFFFAOYSA-N
141268
COC(=O)C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F
C10H3F17O4
MFCD00054723
methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-perfluoropropoxy propoxy propanoate, perfluoro-2,5-dimethyl-3,6-dioxanonanoic acid methyl ester, methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propanoate, methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-heptafluoropropoxy propoxy propanoate, cf3cf2cf2ocf cf3 cf2ocf cf3 cooch3, methyl perfluoro-2,5-dimethyl-3,6-dioxa-nonanoate, methyl 2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoate, 2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoic acid methyl ester, 2-2-heptafluoropropoxy hexafluoropropoxy-2,3,3,3-tetrafluoropropionic acid methyl ester, methyl2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoate mixtureofisomers
methyl 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoate

Specifications

155°C to 156°C
26131-32-8
MFCD00054723
UMADKHFVSXRTGY-UHFFFAOYSA-N
methyl 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoate
141268
97%
1g
C10H3F17O4
methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-perfluoropropoxy propoxy propanoate, perfluoro-2,5-dimethyl-3,6-dioxanonanoic acid methyl ester, methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propanoate, methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-heptafluoropropoxy propoxy propanoate, cf3cf2cf2ocf cf3 cf2ocf cf3 cooch3, methyl perfluoro-2,5-dimethyl-3,6-dioxa-nonanoate, methyl 2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoate, 2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoic acid methyl ester, 2-2-heptafluoropropoxy hexafluoropropoxy-2,3,3,3-tetrafluoropropionic acid methyl ester, methyl2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoate mixtureofisomers
COC(=O)C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F
510.103
510.12
Methyl perfluoro-2,5-dimethyl-3,6-dioxanonanoate