Alfa Aesar™ Methyl perfluoro-2,5-dimethyl-3,6-dioxanonanoate, 97%
Manufacturer:  Alfa Aesar™ L1658606
 View more versions of this product
Catalog No. AAL1658606
Specifications
Specifications
Methyl perfluoro-2,5-dimethyl-3,6-dioxanonanoate
5g
C10H3F17O4
2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoic acid methyl ester
COC(=O)C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F
510.103
510.12
155°C to 156°C
26131-32-8
MFCD00054723
UMADKHFVSXRTGY-UHFFFAOYSA-N
methyl 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoate
141268
97%
Chemical Identifiers
26131-32-8
510.103
UMADKHFVSXRTGY-UHFFFAOYSA-N
141268
COC(=O)C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F
C10H3F17O4
MFCD00054723
2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoic acid methyl ester
methyl 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoate
Documents