Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl pentafluoropropionate, 98%

Chemical Identifiers

Chemical Name or Material Methyl pentafluoropropionate
CAS 378-75-6
Molecular Formula CF3CF2CO2CH3
Formula Weight 178.06
MDL Number MFCD00000430
UN Number UN3272
Beilstein 1777501
Synonym Pentafluoropropionic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1788706

  SDS
alfa aesar™
A1788706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1788714

  SDS
alfa aesar™
A1788714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.06
Chemical Name or Material Methyl pentafluoropropionate
TSCA TSCA
Assay 98%
Molecular Formula CF3CF2CO2CH3
MDL Number MFCD00000430
CAS 378-75-6
UN Number UN3272
Beilstein 1777501
Synonym Pentafluoropropionic acid methyl ester
Health Hazard 1 Danger
Boiling Point 59° to 60°C
Quantity 5g
Flash Point −1°C (30°F)
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
RTECS Number UF6475500
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.