missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl p-tolyl sulfone, 99%

Manufacturer:  Alfa Aesar™ A1153106

 View more versions of this product

Catalog No. AAA1153106



Specifications

Specifications

Methyl p-tolyl sulfone
3185-99-7
C8H10O2S
607296
86°C to 89°C
YYDNBUBMBZRNQQ-UHFFFAOYSA-N
1-methyl-4-methylsulfonylbenzene
18521
5g
99%
MFCD00014742
4-(Methylsulfonyl)toluene
140°C (3mmHg)
CC1=CC=C(C=C1)S(=O)(=O)C
170.226
170.23

Chemical Identifiers

3185-99-7
170.226
YYDNBUBMBZRNQQ-UHFFFAOYSA-N
18521
CC1=CC=C(C=C1)S(=O)(=O)C
C8H10O2S
MFCD00014742
4-(Methylsulfonyl)toluene
1-methyl-4-methylsulfonylbenzene
SDS Documents