Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl p-toluenesulfonylacetate, 98+%

$69.27 - $69.27

Chemical Identifiers

Chemical Name or Material Methyl p-toluenesulfonylacetate
CAS 50397-64-3
Molecular Formula C10H12O4S
Formula Weight 228.27
MDL Number MFCD00025885
Beilstein 2112364
Synonym p-Toluenesulfonylacetic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1314806

 
alfa aesar™
L1314806
5g Each for $69.27
Description & Specifications

Specifications

Formula Weight 228.27
Chemical Name or Material Methyl p-toluenesulfonylacetate
Assay ≥98%
Molecular Formula C10H12O4S
CAS 50397-64-3
Synonym p-Toluenesulfonylacetic acid methyl ester
MDL Number MFCD00025885
Beilstein 2112364
Quantity 5g