Learn More
Methyl p-toluenesulfonate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 170101000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl p-toluenesulfonate | |
80-48-8 | |
292.0°C | |
Authentic | |
Glass bottle | |
1.5162 to 1.5182 | |
MFCD00008417 | |
11, 99 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and benzene | |
CC1=CC=C(C=C1)S(=O)(=O)OC | |
186.23 | |
186.23 |
97% | |
0.2% max. | |
>110°C | |
96% min. (GC) | |
C8H10O3S | |
CH3C6H4SO3CH3 | |
100g | |
methyl p-toluenesulfonate, methyl tosylate, methyl p-tosylate, methyl toluene-4-sulfonate, methyl-p-toluenesulfonate, p-toluenesulfonic acid, methyl ester, benzenesulfonic acid, 4-methyl-, methyl ester, methyl toluene-4-sulphonate, p-toluenesulfonic acid methyl ester, p-methylbenzenesulfonate methyl ester | |
VUQUOGPMUUJORT-UHFFFAOYSA-N | |
methyl 4-methylbenzenesulfonate | |
6645 | |
97% |
Chemical Identifiers
80-48-8 | |
186.23 | |
VUQUOGPMUUJORT-UHFFFAOYSA-N | |
6645 | |
CC1=CC=C(C=C1)S(=O)(=O)OC |
C8H10O3S | |
MFCD00008417 | |
methyl p-toluenesulfonate, methyl tosylate, methyl p-tosylate, methyl toluene-4-sulfonate, methyl-p-toluenesulfonate, p-toluenesulfonic acid, methyl ester, benzenesulfonic acid, 4-methyl-, methyl ester, methyl toluene-4-sulphonate, p-toluenesulfonic acid methyl ester, p-methylbenzenesulfonate methyl ester | |
methyl 4-methylbenzenesulfonate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (
GHS Signal Word: Danger
EINECSNumber : 201-283-5
RTECSNumber : XT7000000
TSCA : TSCA
RUO â Research Use Only