Please login to your online account to display your discounted pricing

Methyl p-toluate, 99%, ACROS Organics™


Quantity 10kg
Packaging Plastic Jerrycan

Manufacturer: thermo fisher scientific  AC414960100

 View more versions of this product

Catalog No. AC414960100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 150.18
Quantity 10kg
Packaging Plastic Jerrycan
EINECS Number 202-784-1
TSCA TSCA
Density 1.0600g/mL
RTECS Number XU2150000
Chemical Name or Material Methyl p-toluate
Assay Percent Range 99%
Linear Formula CH3C6H4COOCH3
MDL Number MFCD00008441
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99%
Molecular Formula C9H10O2
CAS 99-75-2

Specifications

Chemical Name or Material Methyl p-toluate
Name Note 99%
CAS 99-75-2
Assay Percent Range 99%
Molecular Formula C9H10O2
Linear Formula CH3C6H4COOCH3
Formula Weight 150.18
MDL Number MFCD00008441
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.