missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl orotate, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC208781000
Specifications
Methyl orotate | |
Glass bottle | |
244°C | |
6153-44-2 | |
C6H6N2O4 | |
25, 254 | |
4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester, 41c6ifg4mq, 6-carbomethyoxyuracil, methyl 2,6-dihydroxy-4-pyrimidinecarboxylate, methyl 2,6-dihydroxypyrimidine-4-carboxylate, methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate, methyl orotate, orotic acid methyl ester, unii-41c6ifg4mq | |
UUTDWTOZAWFKFW-UHFFFAOYSA-N | |
methyl 2,4-dioxo-1H-pyrimidine-6-carboxylate | |
80257 | |
Powder |
Authentic | |
Brown or White to Yellow | |
100g | |
98% | |
MFCD00010564 | |
15, 6972 | |
Solubility in water: .. Other solubilities: soluble in 1n nh4oh | |
COC(=O)C1=CC(=O)NC(=O)N1 | |
170.12 | |
170.12 | |
98% |
Chemical Identifiers
6153-44-2 | |
170.12 | |
UUTDWTOZAWFKFW-UHFFFAOYSA-N | |
80257 | |
COC(=O)C1=CC(=O)NC(=O)N1 |
C6H6N2O4 | |
MFCD00010564 | |
4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester, 41c6ifg4mq, 6-carbomethyoxyuracil, methyl 2,6-dihydroxy-4-pyrimidinecarboxylate, methyl 2,6-dihydroxypyrimidine-4-carboxylate, methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate, methyl orotate, orotic acid methyl ester, unii-41c6ifg4mq | |
methyl 2,4-dioxo-1H-pyrimidine-6-carboxylate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lens
GHS Signal Word: Warning
EINECSNumber : 228-171-9