missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Orange-Xylene Cyanol, pH 2.9 to 4.6 Lavender to Green, Certified, LabChem™

Manufacturer:  LabChem LC170402

 View more versions of this product

Catalog No. LC170402



Specifications

Specifications

Methyl Orange-Xylene Cyanol
99.8,0.1,0.1
4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
327.34
Certified
Soluble in water
Sulfur compounds; Carbon monoxide; Carbon dioxide
Gray-Green
pH 2.9 to 4.6 Lavender to Green
4463-44-9,547-58-0
Passes Test
Liquid
Poly Bottle
1g/mL
1g/mL
1L
Documents
Promotions

Promotions