Please login to your online account to display your discounted pricing

Methyl Orange, Sodium Salt Grade, ACS Grade, LabChem™

$51.23 - $51.23

Chemical Identifiers

Chemical Name or Material Methyl Orange
CAS 547-58-0
CAS Max % 100
Name Note Sodium Salt Grade
Purity Grade ACS Grade
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
Formula Weight 327.34
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC169808

 
lab chem
LC169808
Amber Glass 25g Each for $51.23
Description & Specifications

Specifications

Formula Weight 327.34
Chemical Name or Material Methyl Orange
Quantity 25g
Name Note Sodium Salt Grade
Purity Grade ACS Grade
Physical Form Powder
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
CAS 547-58-0
UN Number UN2811
Identification Passes Test
Health Hazard 1 Danger
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
Toxic if swallowed.
Health Hazard 3 GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Specific Gravity 1000kg/m3
Recommended Storage Room Temperature
Density 1,000kg/m3
Melting Point 300°C
Color Orange
Safety and Handling

Danger

GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic if swallowed.

Room Temperature