Please login to your online account to display your discounted pricing

Methyl Orange, Modified, Certified, LabChem™

$55.83 - $55.83

Chemical Identifiers

Chemical Name or Material Methyl Orange
CAS 7732-18-5
CAS Max % 99.976
CAS 4463-44-9
CAS Max % 0.014
CAS 547-58-0
CAS Max % 0.01
Name Note Modified
Purity Grade Certified Grade
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC170201

 
lab chem
LC170201
Poly Bottle 500mL Each for $55.83
Description & Specifications

Specifications

Formula Weight 327.34
Purity Grade Certified Grade
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
Identification Passes Test
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
CAS 547-58-0
Physical Form Liquid
CAS Max % 0.01
Name Note Modified
Specific Gravity 1g/mL
Chemical Name or Material Methyl Orange
CAS 4463-44-9
Packaging Poly Bottle
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Quantity 500mL
CAS Max % 99.976
CAS Max % 0.014
Recommended Storage Room Temperature
Density 1g/mL
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Color gray-green
CAS 7732-18-5
Solubility Soluble in water
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature