Please login to your online account to display your discounted pricing

Methyl Orange (Certified ACS), Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  M216500

 View more versions of this product

Catalog No. M216-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Formula Weight 327.33
Melting Point 300°C
Quantity 500g
DOT Information DOT Class 6.1, : Poison
Molecular Formula C14H14N3NaO3S
Color Index No. 13025
Color Orange
Clarity Pass Test
MDL Number MFCD00007502
ChemAlert Storage Symbol Blue
Synonym Helianthin
Packaging Poly Bottle
Visual Transition Interval Pass Test
Chemical Name or Material Methyl Orange
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
CAS 547-58-0

Specifications

Chemical Name or Material Methyl Orange
CAS 547-58-0
Purity Grade Certified ACS Grade
Molecular Formula C14H14N3NaO3S
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
Formula Weight 327.33
MDL Number MFCD00007502
Synonym Helianthin
Accessories
 

SDS & Safety Documents