Please login to your online account to display your discounted pricing

Methyl Orange, 0.1% Aqueous, pH 3.2 to 4.4 Red to Yellow, LabChem™

$28.80 - $28.80

Chemical Identifiers

Chemical Name or Material Methyl Orange
CAS 7732-18-5
CAS Max % 99.9
CAS 547-58-0
CAS Max % 0.1
Name Note 0.1% Aqueous
Purity Grade Certified Grade
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
Formula Weight 327.34
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC170101

 
lab chem
LC170101
Poly Bottle 500mL Each for $28.80
Description & Specifications

Specifications

pH 3.2 to 4.4
Formula Weight 327.34
Purity Grade Certified Grade
Color dark, orange solution
CAS 547-58-0
Recommended Storage Room Temperature
Physical Form Liquid
Packaging Poly Bottle
Identification Passes Test
Solubility Soluble in water
Quantity 500mL
CAS 7732-18-5
Chemical Name or Material Methyl Orange
CAS Max % 0.1
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
CAS Max % 99.9
Name Note 0.1% Aqueous
Decomposition Information Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
Safety and Handling

Room Temperature