Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl o-tolylacetate, 98%

$73.74 - $273.99

Chemical Identifiers

Chemical Name or Material Methyl o-tolylacetate
CAS 40851-62-5
Molecular Formula C10H12O2
Formula Weight 164.21
MDL Number MFCD00082780
Beilstein 2208739
Synonym Methyl 2-methylphenylacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0987706

 
alfa aesar™
L0987706
5g Each for $73.74

AAL0987714

 
alfa aesar™
L0987714
25g Each for $273.99
Description & Specifications

Specifications

Formula Weight 164.21
Chemical Name or Material Methyl o-tolylacetate
MDL Number MFCD00082780
CAS 40851-62-5
Synonym Methyl 2-methylphenylacetate
Assay 98%
Molecular Formula C10H12O2
Beilstein 2208739
Boiling Point 70° to 71°C (1mmHg)
Quantity 5g

Applications
Methyl o-tolylacetate is used as pharmaceutical intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible with oxidizing agents.