missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl o-tolylacetate, 98%

Manufacturer:  Alfa Aesar™ L0987706

 View more versions of this product

Catalog No. AAL0987706



Description

Description

Applications
Methyl o-tolylacetate is used as pharmaceutical intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible with oxidizing agents.
Specifications

Specifications

Methyl o-tolylacetate
5g
C10H12O2
2208739
BLEMRRXGTKTJGT-UHFFFAOYSA-N
methyl 2-(2-methylphenyl)acetate
7021479
98%
70°C to 71°C (1mmHg)
40851-62-5
MFCD00082780
methyl o-tolylacetate, methyl 2-2-methylphenyl acetate, methyl 2-o-tolylacetate, methyl 2-o-tolyl acetate, methyl 2-methylphenylacetate, methyl 2-methylphenyl-acetate, methyl 2-methylphenyl acetate, methyl 2-methyl phenyl acetate, 2-methylbenzeneacetic acid methyl ester, 2-methylphenyl acetic acid methyl ester
CC1=CC=CC=C1CC(=O)OC
164.204
164.21

Chemical Identifiers

40851-62-5
164.204
BLEMRRXGTKTJGT-UHFFFAOYSA-N
7021479
CC1=CC=CC=C1CC(=O)OC
C10H12O2
MFCD00082780
methyl o-tolylacetate, methyl 2-2-methylphenyl acetate, methyl 2-o-tolylacetate, methyl 2-o-tolyl acetate, methyl 2-methylphenylacetate, methyl 2-methylphenyl-acetate, methyl 2-methylphenyl acetate, methyl 2-methyl phenyl acetate, 2-methylbenzeneacetic acid methyl ester, 2-methylphenyl acetic acid methyl ester
methyl 2-(2-methylphenyl)acetate
Documents