missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl O,O'-bis(2,2,2-trifluoroethyl)phosphonoacetate, 90%, ACROS Organics™

$219.22 - $219.22

Chemical Identifiers

CAS 88738-78-7
Molecular Formula C7H9F6O5P
Molecular Weight (g/mol) 318.11
MDL Number MFCD00009901
InChI Key PVSJXEDBEXYLML-UHFFFAOYSA-N
Synonym acetic acid, bis 2,2,2-trifluoroethoxy phosphinyl-, methyl ester
PubChem CID 184894
IUPAC Name methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate
SMILES COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC370610050 SDS View Documents Thermo Fisher Scientific
AC370610050
5g Glass bottle Each for $219.22
 
Description

Description

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

88738-78-7
318.11
PVSJXEDBEXYLML-UHFFFAOYSA-N
184894
COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F
C7H9F6O5P
MFCD00009901
acetic acid, bis 2,2,2-trifluoroethoxy phosphinyl-, methyl ester
methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate

Specifications

1.5040g/mL
Authentic
1.3700 to 1.3740
Brown-Orange to Orange
88738-78-7
C7H9F6O5P
MFCD00009901
acetic acid, bis 2,2,2-trifluoroethoxy phosphinyl-, methyl ester
COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F
318.11
318.11
90%
>110°C
Glass bottle
1.504
5g
88% min. (GC)
CH3O2CCH2P(O)(OCH2CF3)2
12,320; 13,186
PVSJXEDBEXYLML-UHFFFAOYSA-N
methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate
184894
Liquid
Methyl O,O'-bis(2,2,2-trifluoroethyl)phosphonoacetate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning