Learn More
Methyl nonanoate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 385561000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl nonanoate | |
0.8740g/mL | |
81°C | |
94% min. (GC) | |
C10H20O2 | |
CH3(CH2)7C(O)OCH3 | |
100g | |
methyl pelargonate, pelargonic acid methyl ester, methyl nonylate, nonanoic acid, methyl ester, methyl n-nonanoate, nonanoic acid methyl ester, unii-7xkd6qoh68, fema no. 2724, methyl ester nonanoic acid, 7xkd6qoh68 | |
IJXHLVMUNBOGRR-UHFFFAOYSA-N | |
methyl nonanoate | |
15606 | |
172.27 |
1731-84-6 | |
213°C | |
Authentic | |
Glass bottle | |
1.4210 to 1.423 | |
MFCD00009569 | |
0.874 | |
Solubility in water: practically insoluble | |
CCCCCCCCC(=O)OC | |
172.27 | |
CHEBI:44499 | |
95% |
Chemical Identifiers
1731-84-6 | |
172.27 | |
IJXHLVMUNBOGRR-UHFFFAOYSA-N | |
15606 | |
methyl nonanoate |
C10H20O2 | |
MFCD00009569 | |
methyl pelargonate, pelargonic acid methyl ester, methyl nonylate, nonanoic acid, methyl ester, methyl n-nonanoate, nonanoic acid methyl ester, unii-7xkd6qoh68, fema no. 2724, methyl ester nonanoic acid, 7xkd6qoh68 | |
CHEBI:44499 | |
CCCCCCCCC(=O)OC |
Safety and Handling
GHS H Statement:
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 217-052-7
RUO â Research Use Only