missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Nonafluorobutyl Ketone, TCI America™

$368.20 - $368.20

Chemical Identifiers

CAS 678-18-2
Molecular Formula C6H3F9O
Molecular Weight (g/mol) 262.075
MDL Number MFCD00153209
InChI Key LPPFRPMJUNOKRS-UHFFFAOYSA-N
Synonym 1H,1H,1H-Nonafluoro-2-hexanone, Methyl Perfluorobutyl Ketone, 3,3,4,4,5,5,6,6,6-Nonafluoro-2-hexanone
PubChem CID 2775461
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohexan-2-one
SMILES CC(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N06455G View Documents TCI America
N06455G
5g Each for $368.20
 
Specifications

Chemical Identifiers

678-18-2
262.075
LPPFRPMJUNOKRS-UHFFFAOYSA-N
2775461
CC(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F
C6H3F9O
MFCD00153209
1H,1H,1H-Nonafluoro-2-hexanone, Methyl Perfluorobutyl Ketone, 3,3,4,4,5,5,6,6,6-Nonafluoro-2-hexanone
3,3,4,4,5,5,6,6,6-nonafluorohexan-2-one

Specifications

69°C
5g
C6H3F9O
1224
LPPFRPMJUNOKRS-UHFFFAOYSA-N
3,3,4,4,5,5,6,6,6-nonafluorohexan-2-one
2775461
Clear Liquid at 20°C
Methyl Nonafluorobutyl Ketone
Colorless
678-18-2
MFCD00153209
1H,1H,1H-Nonafluoro-2-hexanone, Methyl Perfluorobutyl Ketone, 3,3,4,4,5,5,6,6,6-Nonafluoro-2-hexanone
CC(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F
262.075
262.07
≥99.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No