missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Nonadecafluorodecanoate 97.0+%, TCI America™

$85.58 - $255.11

Chemical Identifiers

CAS 307-79-9
Molecular Formula C11H3F19O2
Molecular Weight (g/mol) 528.113
MDL Number MFCD00153211
InChI Key QJFHNYDPNSFJMR-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoic acid methyl ester, acmc-1cjo4, decanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluoro-, methyl ester, methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecakis fluoranyl decanoate, methyl nonadecafluorodecanoate, methyl perfluorodecanoate, nonadecafluorodecanoic acid methyl ester, nonadecafluorodecanoic acid methyl ester; perfluorodecanoic acid methyl ester, perfluorodecanoic acid methyl ester
PubChem CID 2733270
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoate
SMILES COC(=O)C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19165G View Documents Tci America
M19165G
5g Each for $85.58
 
M191625G View Documents Tci America
M191625G
25g Each for $255.11
 
Specifications

Chemical Identifiers

307-79-9
528.113
QJFHNYDPNSFJMR-UHFFFAOYSA-N
2733270
COC(=O)C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C11H3F19O2
MFCD00153211
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoic acid methyl ester, acmc-1cjo4, decanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluoro-, methyl ester, methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecakis fluoranyl decanoate, methyl nonadecafluorodecanoate, methyl perfluorodecanoate, nonadecafluorodecanoic acid methyl ester, nonadecafluorodecanoic acid methyl ester; perfluorodecanoic acid methyl ester, perfluorodecanoic acid methyl ester
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoate

Specifications

193°C
5g
C11H3F19O2
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoic acid methyl ester, acmc-1cjo4, decanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluoro-, methyl ester, methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecakis fluoranyl decanoate, methyl nonadecafluorodecanoate, methyl perfluorodecanoate, nonadecafluorodecanoic acid methyl ester, nonadecafluorodecanoic acid methyl ester; perfluorodecanoic acid methyl ester, perfluorodecanoic acid methyl ester
COC(=O)C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
528.113
528.11
≥97.0% (GC)
Yellow
307-79-9
MFCD00153211
QJFHNYDPNSFJMR-UHFFFAOYSA-N
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoate
2733270
Clear Liquid at 20°C
Methyl Nonadecafluorodecanoate
Safety and Handling

Safety and Handling

TSCA : No