Please login to your online account to display your discounted pricing

Methyl nonadecafluorodecanoate, 94%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl nonadecafluorodecanoate
Name Note 94%
CAS 307-79-9
Assay ≥92%
Assay Percent Range 94%
Molecular Formula C11H3F19O2
Linear Formula CF3(CF)8COCH3
Formula Weight 528.11
MDL Number MFCD00153211
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC342230250

  SDS
thermo fisher scientific
AC342230250
Glass Bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 528.11
Assay ≥92%
Quantity 25mL
Color Clear colorless
Density 1.76g/mL
Infrared Spectrum Authentic
Refractive Index 1.3070 to 1.3090 (20°C, 589nm)
Packaging Glass Bottle
Molecular Formula C11H3F19O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material Methyl nonadecafluorodecanoate
Name Note 94%
MDL Number MFCD00153211
CAS 307-79-9
Assay Percent Range 94%
Linear Formula CF3(CF)8COCH3
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.