Please login to your online account to display your discounted pricing

Methyl nonadecafluorodecanoate, 94%, ACROS Organics™


Quantity 25mL
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC342230250

 View more versions of this product

Catalog No. AC342230250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 528.11
Assay ≥92%
Quantity 25mL
Color Clear colorless
Density 1.76g/mL
Infrared Spectrum Authentic
Refractive Index 1.3070 to 1.3090 (20°C, 589nm)
Packaging Glass Bottle
Molecular Formula C11H3F19O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material Methyl nonadecafluorodecanoate
Name Note 94%
MDL Number MFCD00153211
CAS 307-79-9
Assay Percent Range 94%
Linear Formula CF3(CF)8COCH3

Specifications

Chemical Name or Material Methyl nonadecafluorodecanoate
Name Note 94%
CAS 307-79-9
Assay ≥92%
Assay Percent Range 94%
Molecular Formula C11H3F19O2
Linear Formula CF3(CF)8COCH3
Formula Weight 528.11
MDL Number MFCD00153211
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents