Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl N-trimethylsilylcarbamate, 96%

$62.84 - $62.84

Chemical Identifiers

Chemical Name or Material Methyl N-trimethylsilylcarbamate
CAS 18147-09-6
Molecular Formula C5H13NO2Si
Linear Formula (CH3)3SiNHCO2CH3
Formula Weight 147.25
MDL Number MFCD00236515
Beilstein 2350760
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1735206

  SDS
alfa aesar™
L1735206
5g Each for $62.84
Description & Specifications

Specifications

Formula Weight 147.25
Chemical Name or Material Methyl N-trimethylsilylcarbamate
Assay 96%
CAS 18147-09-6
Beilstein 2350760
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C5H13NO2Si
Linear Formula (CH3)3SiNHCO2CH3
MDL Number MFCD00236515
Boiling Point 66° to 67°C (15mmHg)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.