missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl N-(tert-Butoxycarbonyl)-L-pyroglutamate 98.0+%, TCI America™
$114.48 - $394.84
Chemical Identifiers
CAS | 108963-96-8 |
---|---|
Molecular Formula | C11H17NO5 |
Molecular Weight (g/mol) | 243.259 |
MDL Number | MFCD06809720 |
InChI Key | FNTAOUUEQHKLIU-ZETCQYMHSA-N |
Synonym | boc-l-pyroglutamic acid methyl ester, boc-pyr-ome, s-1-tert-butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate, methyl l-n-boc-pyroglutamate, 1-tert-butyl 2-methyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, methyl boc-l-pyroglutamate, s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 1-tert-butyl 2-methyl s-5-oxopyrrolidine-1,2-dicarboxylate, boc-pglu-ome, pubchem12135 |
PubChem CID | 10868485 |
IUPAC Name | 1-O-tert-butyl 2-O-methyl (2S)-5-oxopyrrolidine-1,2-dicarboxylate |
SMILES | CC(C)(C)OC(=O)N1C(CCC1=O)C(=O)OC |
Chemical Identifiers
108963-96-8 | |
243.259 | |
FNTAOUUEQHKLIU-ZETCQYMHSA-N | |
10868485 | |
CC(C)(C)OC(=O)N1C(CCC1=O)C(=O)OC |
C11H17NO5 | |
MFCD06809720 | |
boc-l-pyroglutamic acid methyl ester, boc-pyr-ome, s-1-tert-butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate, methyl l-n-boc-pyroglutamate, 1-tert-butyl 2-methyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, methyl boc-l-pyroglutamate, s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 1-tert-butyl 2-methyl s-5-oxopyrrolidine-1,2-dicarboxylate, boc-pglu-ome, pubchem12135 | |
1-O-tert-butyl 2-O-methyl (2S)-5-oxopyrrolidine-1,2-dicarboxylate |
Specifications
108963-96-8 | |
White | |
MFCD06809720 | |
boc-l-pyroglutamic acid methyl ester, boc-pyr-ome, s-1-tert-butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate, methyl l-n-boc-pyroglutamate, 1-tert-butyl 2-methyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, methyl boc-l-pyroglutamate, s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 1-tert-butyl 2-methyl s-5-oxopyrrolidine-1,2-dicarboxylate, boc-pglu-ome, pubchem12135 | |
CC(C)(C)OC(=O)N1C(CCC1=O)C(=O)OC | |
243.259 | |
243.26 | |
Crystalline Powder |
71°C | |
C11H17NO5 | |
5 g | |
FNTAOUUEQHKLIU-ZETCQYMHSA-N | |
1-O-tert-butyl 2-O-methyl (2S)-5-oxopyrrolidine-1,2-dicarboxylate | |
10868485 | |
≥98.0% (GC) | |
Methyl N-(tert-Butoxycarbonyl)-L-pyroglutamate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator