Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl n-propyl sulfide, 99%

Chemical Identifiers

Chemical Name or Material Methyl n-propyl sulfide
CAS 3877-15-4
Molecular Formula CH3(CH2)2SCH3
Formula Weight 90.19
MDL Number MFCD00015219
UN Number UN1993
Beilstein 1730904
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0934609

 
alfa aesar™
L0934609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0934618

 
alfa aesar™
L0934618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 90.19
Chemical Name or Material Methyl n-propyl sulfide
Assay 99%
Molecular Formula CH3(CH2)2SCH3
MDL Number MFCD00015219
CAS 3877-15-4
UN Number UN1993
Beilstein 1730904
Health Hazard 1 Danger
Quantity 10g
Melting Point −113°C
Boiling Point 94° to 96°C
Flash Point 11°C (51°F)
Odor Stench
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.