missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Myristate, TCI America™

Manufacturer:  TCI America M0482500ML

 View more versions of this product

Catalog No. M0482500ML



Specifications

Specifications

Methyl Myristate
Yellow
124-10-7
MFCD00008983
ZAZKJZBWRNNLDS-UHFFFAOYSA-N
methyl tetradecanoate
31284
Clear Liquid at 20°C
173°C
500mL
C15H30O2
methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci
CCCCCCCCCCCCCC(=O)OC
242.403
242.40
≥98.0% (GC)

Chemical Identifiers

124-10-7
242.403
ZAZKJZBWRNNLDS-UHFFFAOYSA-N
31284
CCCCCCCCCCCCCC(=O)OC
C15H30O2
MFCD00008983
methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci
methyl tetradecanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

TSCA : Yes

Documents