Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl (methylthio)methyl sulfoxide, 97%

$75.95 - $75.95

Chemical Identifiers

Chemical Name or Material Methyl (methylthio)methyl sulfoxide
CAS 33577-16-1
Molecular Formula C3H8OS2
Formula Weight 124.22
MDL Number MFCD00002091
Beilstein 906789
Synonym Formaldehyde dimethyl thioacetal monoxide; Methyl (methylsulfinyl)methyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0051009

  SDS
alfa aesar™
L0051009
10g Each for $75.95
Description & Specifications

Specifications

Formula Weight 124.22
MDL Number MFCD00002091
CAS 33577-16-1
Assay 97%
Molecular Formula C3H8OS2
Synonym Formaldehyde dimethyl thioacetal monoxide; Methyl (methylsulfinyl)methyl sulfide
Boiling Point 222° to 226°C
Quantity 10g
Flash Point >110°C (230°F)
TSCA TSCA
Beilstein 906789
Chemical Name or Material Methyl (methylthio)methyl sulfoxide