missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl (Methylthio)acetate, TCI America™

$47.36 - $187.92

Chemical Identifiers

CAS 16630-66-3
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00039809
InChI Key HZYCAKGEXXKCDM-UHFFFAOYSA-N
Synonym methyl methylthio acetate, methyl 2-methylthio acetate, methyl 2-methylsulfanyl acetate, acetic acid, methylthio-, methyl ester, methyl methylithio acetate, unii-gn58c28ml9, methyl methylthioacetate, acmc-1c4wc, methyl methylthio acetate, methyl 3-methylthio acetate
PubChem CID 85522
IUPAC Name methyl 2-methylsulfanylacetate
SMILES COC(=O)CSC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M22965G View Documents TCI America
M22965G
5g Each for $47.36
 
M229625G View Documents TCI America
M229625G
25g Each for $187.92
 
Specifications

Chemical Identifiers

16630-66-3
120.166
HZYCAKGEXXKCDM-UHFFFAOYSA-N
85522
COC(=O)CSC
C4H8O2S
MFCD00039809
methyl methylthio acetate, methyl 2-methylthio acetate, methyl 2-methylsulfanyl acetate, acetic acid, methylthio-, methyl ester, methyl methylithio acetate, unii-gn58c28ml9, methyl methylthioacetate, acmc-1c4wc, methyl methylthio acetate, methyl 3-methylthio acetate
methyl 2-methylsulfanylacetate

Specifications

60°C
5g
C4H8O2S
3272
HZYCAKGEXXKCDM-UHFFFAOYSA-N
methyl 2-methylsulfanylacetate
85522
Clear Liquid at 20°C
Methyl (Methylthio)acetate
Yellow
16630-66-3
MFCD00039809
methyl methylthio acetate, methyl 2-methylthio acetate, methyl 2-methylsulfanyl acetate, acetic acid, methylthio-, methyl ester, methyl methylithio acetate, unii-gn58c28ml9, methyl methylthioacetate, acmc-1c4wc, methyl methylthio acetate, methyl 3-methylthio acetate
COC(=O)CSC
120.166
120.17
≥99.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator