missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl (Methylsulfinyl)methyl Sulfide, TCI America™

Manufacturer:  TCI America M080525G

 View more versions of this product

Catalog No. M080525G



Specifications

Specifications

Methyl (Methylsulfinyl)methyl Sulfide
Yellow
33577-16-1
MFCD00002091
OTKFCIVOVKCFHR-UHFFFAOYSA-N
methylsulfanyl(methylsulfinyl)methane
99129
Clear Liquid at 20°C
93°C
25g
C3H8OS2
methyl methylsulfinylmethyl sulfide, methyl methylsulfinyl methyl sulfide, methane, methylsulfinyl methylthio, methyl methylthio methyl sulfoxide, famso, methanesulfinyl-methylsulfanyl-methane, formaldehyde dimethyl mercaptal s-oxide, methylsulfinyl methylthio methane, methyl methylthiomethyl sulphoxide, formaldehyde methyl mercaptal s-oxide
CSCS(=O)C
124.216
124.22
≥96.0% (GC)

Chemical Identifiers

33577-16-1
124.216
OTKFCIVOVKCFHR-UHFFFAOYSA-N
99129
CSCS(=O)C
C3H8OS2
MFCD00002091
methyl methylsulfinylmethyl sulfide, methyl methylsulfinyl methyl sulfide, methane, methylsulfinyl methylthio, methyl methylthio methyl sulfoxide, famso, methanesulfinyl-methylsulfanyl-methane, formaldehyde dimethyl mercaptal s-oxide, methylsulfinyl methylthio methane, methyl methylthiomethyl sulphoxide, formaldehyde methyl mercaptal s-oxide
methylsulfanyl(methylsulfinyl)methane
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2801

TSCA : Yes

Documents