missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl (Methylsulfinyl)methyl Sulfide, TCI America™

$62.64 - $183.34

Chemical Identifiers

CAS 33577-16-1
Molecular Formula C3H8OS2
Molecular Weight (g/mol) 124.216
MDL Number MFCD00002091
InChI Key OTKFCIVOVKCFHR-UHFFFAOYSA-N
Synonym methyl methylsulfinylmethyl sulfide, methyl methylsulfinyl methyl sulfide, methane, methylsulfinyl methylthio, methyl methylthio methyl sulfoxide, famso, methanesulfinyl-methylsulfanyl-methane, formaldehyde dimethyl mercaptal s-oxide, methylsulfinyl methylthio methane, methyl methylthiomethyl sulphoxide, formaldehyde methyl mercaptal s-oxide
PubChem CID 99129
IUPAC Name methylsulfanyl(methylsulfinyl)methane
SMILES CSCS(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M08055G View Documents TCI America
M08055G
5g Each for $62.64
 
M080525G View Documents TCI America
M080525G
25g Each for $183.34
 
Specifications

Chemical Identifiers

33577-16-1
124.216
OTKFCIVOVKCFHR-UHFFFAOYSA-N
99129
CSCS(=O)C
C3H8OS2
MFCD00002091
methyl methylsulfinylmethyl sulfide, methyl methylsulfinyl methyl sulfide, methane, methylsulfinyl methylthio, methyl methylthio methyl sulfoxide, famso, methanesulfinyl-methylsulfanyl-methane, formaldehyde dimethyl mercaptal s-oxide, methylsulfinyl methylthio methane, methyl methylthiomethyl sulphoxide, formaldehyde methyl mercaptal s-oxide
methylsulfanyl(methylsulfinyl)methane

Specifications

93°C
5g
C3H8OS2
methyl methylsulfinylmethyl sulfide, methyl methylsulfinyl methyl sulfide, methane, methylsulfinyl methylthio, methyl methylthio methyl sulfoxide, famso, methanesulfinyl-methylsulfanyl-methane, formaldehyde dimethyl mercaptal s-oxide, methylsulfinyl methylthio methane, methyl methylthiomethyl sulphoxide, formaldehyde methyl mercaptal s-oxide
CSCS(=O)C
124.216
124.22
≥96.0% (GC)
Yellow
33577-16-1
MFCD00002091
OTKFCIVOVKCFHR-UHFFFAOYSA-N
methylsulfanyl(methylsulfinyl)methane
99129
Clear Liquid at 20°C
Methyl (Methylsulfinyl)methyl Sulfide
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2801

TSCA : Yes