Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl methylphenylphosphinate, 98%

$156.70 - $616.27

Chemical Identifiers

Chemical Name or Material Methyl methylphenylphosphinate
CAS 6389-79-3
Molecular Formula C8H11O2P
Formula Weight 170.15
MDL Number MFCD00013909
Synonym Methylphenylphosphinic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1876014

  SDS
alfa aesar™
A1876014
25g Each for $616.27

AAA1876006

  SDS
alfa aesar™
A1876006
5g Each for $156.70

AAA1876003

  SDS
alfa aesar™
A1876003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Methyl methylphenylphosphinate
CAS 6389-79-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C8H11O2P
Formula Weight 170.15
MDL Number MFCD00013909
Synonym Methylphenylphosphinic acid methyl ester
Boiling Point 148° to 149°C (17mmHg)
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.