missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl methoxyacetate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC165090250

 View more versions of this product

Catalog No. AC165090250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyl methoxyacetate
35°C
Glass bottle
1.05
Colorless to Brown
99%
99%
CH3OCH2CO2CH3
03, 236
Solubility in water: soluble
COCC(=O)OC
104.11
CHEBI:34841
Liquid
1.0500g/mL
Authentic
1.3954 to 1.3974
129°C to 130°C
25g
6290-49-9
C4H8O3
MFCD00008451
acetic acid, 2-methoxy-, methyl ester
QRMHDGWGLNLHMN-UHFFFAOYSA-N
methyl 2-methoxyacetate
80507
104.11
99%

Chemical Identifiers

6290-49-9
104.11
QRMHDGWGLNLHMN-UHFFFAOYSA-N
80507
methyl 2-methoxyacetate
C4H8O3
MFCD00008451
acetic acid, 2-methoxy-, methyl ester
CHEBI:34841
COCC(=O)OC
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-539-9

RTECSNumber : AI8910000

SDS Documents